Invitrogen
Manufacturer Code:PA5101498
Catalog # PIPA5101498
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CPN1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 0610011F20Rik ACBP Anaphylatoxin inactivator arginine carboxypeptidase carboxypeptidase K carboxypeptidase N catalytic chain carboxypeptidase N catalytic subunit Carboxypeptidase N polypeptide 1 carboxypeptidase N polypeptide 1 50 kD carboxypeptidase N small subunit carboxypeptidase N subunit 1 carboxypeptidase N polypeptide 1 carboxypeptidase N polypeptide 1 50kD CPN Cpn1 FLJ40792 kininase I kininase-1 Lysine carboxypeptidase plasma carboxypeptidase B SCPN Serum carboxypeptidase N; Anaphylatoxin inactivator; Arginine carboxypeptidase; carboxypeptidase K; Carboxypeptidase N catalytic chain; carboxypeptidase N catalytic subunit; Carboxypeptidase N polypeptide 1; carboxypeptidase N polypeptide 1 50 kD; Carboxypeptidase N small subunit; carboxypeptidase N, polypeptide 1; carboxypeptidase N, polypeptide 1, 50kD; CPN; kininase I; Kininase-1; Lysine carboxypeptidase; Plasma carboxypeptidase B; SCPN; Serum carboxypeptidase N
Gene Aliases: 0610011F20Rik; ACBP; CPN; CPN1; SCPN
UniProt ID: (Human) P15169, (Mouse) Q9JJN5
Entrez Gene ID: (Human) 1369, (Mouse) 93721
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.