Abnova
Manufacturer Code:PAB17651
Catalog # 89-119-316
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of human CPN1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of CPN1.
For Research Use Only
Protein Aliases: Anaphylatoxin inactivator; Arginine carboxypeptidase; carboxypeptidase K; Carboxypeptidase N catalytic chain; carboxypeptidase N catalytic subunit; Carboxypeptidase N polypeptide 1; carboxypeptidase N polypeptide 1 50 kD; Carboxypeptidase N small subunit; carboxypeptidase N, polypeptide 1; CPN; FLJ40792; kininase I; Kininase-1; Lysine carboxypeptidase; Plasma carboxypeptidase B; SCPN; Serum carboxypeptidase N
Gene Aliases: ACBP; CPN; CPN1; SCPN
UniProt ID: (Human) P15169
Entrez Gene ID: (Human) 1369
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.