Invitrogen
Manufacturer Code:PA532059
Catalog # PIPA532059
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 158 of Human Complexin 3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Complexin III; Complexin-3; CPX III; CPX-III CPXIII CPX III complexin III complexin-3; lectin, mannose-binding, 1 like
Gene Aliases: BC016632; Cplx3; CpxIII; ERGIC-53L; ERGL; Lamn1l; Lman1l
UniProt ID: (Mouse) Q8R1B5
Entrez Gene ID: (Mouse) 235415
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.