Novus Biologicals
Manufacturer Code:NBP300305AF405
Catalog # NB065479
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant CPLX3 (Q8WVH0 Met1-Lys154). |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: complexin 3 Complexin III complexin-3 CPX III CPXIII CPX-III DKFZp434E1719 FLJ00167 FLJ00250 FLJ13993; Complexin III; Complexin-3; CPX III
Gene Aliases: CPLX3; CPX-III; CPXIII; Nbla11589
UniProt ID: (Human) Q8WVH0
Entrez Gene ID: (Human) 594855
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.