Invitrogen
Manufacturer Code:MA529186
Catalog # PIMA529186
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 21 |
Immunogen | Recombit Human Complexin-2/CPLX2 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Complexin II; Complexin-2; CPLX2 CPX2 CPX-II Hfb1 Synaphin 1 complexin 2 Complexin II complexin-2 CPX II CPX2 CPX-2 DKFZp547D155 Hfb1 MGC138492 synaphin 1921-L synaphin-1 Synaphin-1; CPX II; synaphin 1; Synaphin-1
Gene Aliases: 921-L; CPLX2; CPX-2; CPX2; Hfb1
UniProt ID: (Human) Q6PUV4
Entrez Gene ID: (Human) 10814
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.