Invitrogen
Manufacturer Code:PA569824
Catalog # PIPA569824
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of human CPLX2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 921-L; Complexin II; Complexin-2; CPLX2 CPX2 CPX-II Hfb1 Synaphin 1 complexin 2 Complexin II complexin-2 CPX II CPX2 CPX-2 DKFZp547D155 Hfb1 MGC138492 synaphin 1921-L synaphin-1 Synaphin-1; CPX II; synaphin 1; Synaphin-1
Gene Aliases: 921-L; AI413745; AW492120; CPLX2; CPX-2; CPX2; Hfb1; wu:fc27b12
UniProt ID: (Human) Q6PUV4, (Bovine) P84088, (Rat) P84087, (Mouse) P84086
Entrez Gene ID: (Human) 10814, (Rabbit) 100339715, (Dog) 489107, (Horse) 100058774, (Bovine) 281711, (Guinea pig) 100730004, (Rat) 116657, (Mouse) 12890, (Zebrafish) 556701
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.