Invitrogen
Manufacturer Code:PA5113049
Catalog # PIPA5113049
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Complexin-2 protein (1-134AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 921-L AI413745 AW492120 BOS_10122 complexin 2 Complexin II Complexin-2 Cplx2 CPX II CPX2 CPX-2 CPX-II DKFZp547D155 Hfb1 MGC138492 synaphin 1 Synaphin-1 wu:fc27b12; Complexin II; Complexin-2; CPX II; synaphin 1; Synaphin-1
Gene Aliases: 921-L; CPLX2; CPX-2; CPX2; Hfb1
UniProt ID: (Human) Q6PUV4, (Rat) P84087
Entrez Gene ID: (Human) 10814, (Rat) 116657
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.