Invitrogen
Manufacturer Code:PA5106398
Catalog # PIPA5106398
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CPLX2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 921-L; 921-L AI413745 AW492120 complexin 2 complexin II Complexin-2 CPLX2 CPX II CPX2 CPX-2 CPX-II DKFZp547D155 Hfb1 MGC138492 synaphin 1 synaphin-1; Complexin II; Complexin-2; CPX II; synaphin 1; Synaphin-1
Gene Aliases: 921-L; AI413745; AW492120; CPLX2; CPX-2; CPX2; Hfb1
UniProt ID: (Human) Q6PUV4, (Rat) P84087, (Mouse) P84086
Entrez Gene ID: (Human) 10814, (Rat) 116657, (Mouse) 12890
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.