Proteintech
Manufacturer Code:102462AP150UL
Catalog # 50-172-6146
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CPLX1 Fusion Protein Ag0387 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 921-S; complexin 1 Complexin I complexin1 CPLX1 CPX I CPX1 Synaphin 2; Complexin I; Complexin-1; CPX I; synaphin 2; Synaphin-2
Gene Aliases: 921-S; CPLX1; CPX-I; CPX1
UniProt ID: (Human) O14810, (Rat) P63041, (Mouse) P63040
Entrez Gene ID: (Human) 10815, (Rat) 64832, (Mouse) 12889
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.