Invitrogen
Manufacturer Code:PA579858
Catalog # PIPA579858
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human CPI17 alpha recombit protein (Position: L30-Q126). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 17 kDa PKC-potentiated inhibitory protein of PP1; 17-kDa PKC-potentiated inhibitory protein of PP1; 17-KDa protein; CPI-17; PKC-potentiated inhibitory protein of PP1; Protein kinase C-potentiated inhibitor protein of 17 kDa; Protein phosphatase 1 regulatory subunit 14A; Protein phosphatase 1 regulatory subunit 14A 17 kDa PKC-potentiated inhibitory protein of PP1 Protein kinase C-potentiated inhibitor protein of 17 kDa CPI-17 PPP1R14A CPI17 PPP1INL; protein phosphatase 1, regulatory (inhibitor) subunit 14A
Gene Aliases: 1110001M11Rik; Cpi; CPI-17; CPI17; PPP1INL; PPP1R14A
UniProt ID: (Human) Q96A00, (Rat) Q99MC0, (Mouse) Q91VC7
Entrez Gene ID: (Human) 94274, (Rat) 114004, (Mouse) 68458
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.