Proteintech
Manufacturer Code:253421AP150UL
Catalog # 50-173-4863
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CPE binding protein 4 CPE BP4 CPEB4 hCPEB 4 KIAA1673; CPE-binding protein 4; CPE-BP4; Cytoplasmic polyadenylation element-binding protein 4
Gene Aliases: CPE-BP4; Cpeb-4; CPEB4; hCPEB-4; KIAA1673; mCPEB-4
UniProt ID: (Human) Q17RY0, (Mouse) Q7TN98
Entrez Gene ID: (Human) 80315, (Mouse) 67579, (Rat) 303010
Molecular Function:
RNA binding protein
mRNA polyadenylation factor
mRNA processing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.