Invitrogen
Manufacturer Code:PA544273
Catalog # PIPA544273
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human CPEB3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CPE-binding protein 3; CPE-BP3; cytoplasmic polyadenylation element binding protein 3; Cytoplasmic polyadenylation element-binding protein 3; hCPEB-3
Gene Aliases: 4831444O18Rik; CPE-BP3; CPEB3; KIAA0940; mKIAA0940; RGD1564670
UniProt ID: (Human) Q8NE35, (Mouse) Q7TN99
Entrez Gene ID: (Human) 22849, (Bovine) 537016, (Dog) 486801, (Horse) 100061932, (Rabbit) 100341373, (Pig) 100152117, (Rat) 309510, (Mouse) 208922
Molecular Function: RNA binding protein mRNA polyadenylation factor mRNA processing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.