Proteintech
Manufacturer Code:127801AP150UL
Catalog # 50-557-079
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CP110 Fusion Protein Ag3489 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: centriolar coiled coil protein 110kDa; centriolar coiled-coil protein 110kDa; Centriolar coiled-coil protein of 110 kDa; centrosomal protein CP110; Centrosomal protein of 110 kDa; Centrosomal protein of 110 kDa CP110 CP110 protein KIAA0419; Cep110; CP110
Gene Aliases: 6330503K22Rik; AA415922; AI427129; AW557948; CCP110; CEP110; CP110; KIAA0419; RGD1310509
UniProt ID: (Human) O43303, (Mouse) Q7TSH4
Entrez Gene ID: (Human) 9738, (Dog) 479826, (Mouse) 101565, (Rat) 361634
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.