Proteintech
Manufacturer Code:257521AP150UL
Catalog # 50-173-5055
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | COX20 Fusion Protein Ag22520 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: COX20 Cox2 chaperone homolog; COX20 cytochrome c oxidase assembly factor; Cytochrome c oxidase assembly protein COX20, mitochondrial; FAM36A Protein FAM36A; family with sequence similarity 36, member A
Gene Aliases: COX20; FAM36A
UniProt ID: (Human) Q5RI15
Entrez Gene ID: (Human) 116228
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.