Novus Biologicals
Manufacturer Code:NBP299469IR
Catalog # NB085270
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human COX17. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: COX17 (yeast) homolog cytochrome c oxidase assembly protein COX17 cytochrome c oxidase assembly homolog (S. cerevisiae) cytochrome c oxidase assembly protein (S. cerevisiae) cytochrome c oxidase assembly protein (yeast) cytochrome c oxidase copper chaperone human homolog of yeast mitochondrial copper recruitment MGC104397 MGC117386; COX17 cytochrome c oxidase assembly homolog; COX17 cytochrome c oxidase copper chaperone; cytochrome c oxidase 17 copper chaperone; cytochrome c oxidase assembly homolog 17; Cytochrome c oxidase copper chaperone; human homolog of yeast mitochondrial copper recruitment
Gene Aliases: COX17
UniProt ID: (Human) Q14061
Entrez Gene ID: (Human) 10063
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.