Invitrogen
Manufacturer Code:PA530253
Catalog # PIPA530253
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 178 and 410 of Human COX15 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: COX15 cytochrome c oxidase assembly homolog; COX15 homolog, cytochrome c oxidase assembly protein; cytochrome c oxidase assembly homolog 15; Cytochrome c oxidase assembly protein COX15 homolog; cytochrome c oxidase assembly protein COX15 homolog cytochrome c oxidase subunit 15; cytochrome c oxidase subunit 15
Gene Aliases: CEMCOX2; COX15
UniProt ID: (Human) Q7KZN9
Entrez Gene ID: (Human) 1355
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.