Proteintech
Manufacturer Code:114981AP150UL
Catalog # 50-172-7212
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | COX11 Fusion Protein Ag2033 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: COX11 COX11P; COX11 cytochrome c oxidase copper chaperone; COX11 homolog, cytochrome c oxidase assembly protein; cytochrome c oxidase assembly homolog 11; cytochrome c oxidase assembly protein 11; Cytochrome c oxidase assembly protein COX11, mitochondrial; cytochrome c oxidase subunit 11
Gene Aliases: 2010004I09Rik; COX11; COX11P
UniProt ID: (Human) Q9Y6N1, (Mouse) Q6P8I6
Entrez Gene ID: (Human) 1353, (Mouse) 69802, (Rat) 690300
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.