Invitrogen
Manufacturer Code:PA577244
Catalog # PIPA577244
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human COX10. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: COX10 heme A:farnesyltransferase cytochrome c oxidase assembly factor; COX10 homolog, cytochrome c oxidase assembly protein, heme A: farnesyltransferase; cytochrome c oxidase assembly homolog 10; cytochrome c oxidase assembly protein; cytochrome c oxidase subunit X; heme A: farnesyltransferase; Heme O synthase; Protoheme IX farnesyltransferase mitochondrial Heme O synthase COX10; Protoheme IX farnesyltransferase, mitochondrial; similar to COX10 homolog, cytochrome c oxidase assembly protein, heme A: farnesyltransferase
Gene Aliases: 2410004F01Rik; AU042636; COX10
UniProt ID: (Human) Q12887, (Mouse) Q8CFY5
Entrez Gene ID: (Human) 1352, (Mouse) 70383, (Rat) 691853
Molecular Function: acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.