Invitrogen
Manufacturer Code:PA5115045
Catalog # PIPA5115045
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human COX10 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2410004F01Rik AU042636 Cox10 COX10 heme A:farnesyltransferase cytochrome c oxidase assembly factor COX10 homolog cytochrome c oxidase assembly protein heme A: farnesyltransferase COX10 heme A:farnesyltransferase cytochrome c oxidase assembly factor cytochrome c oxidase assembly homolog 10 cytochrome c oxidase assembly protein cytochrome c oxidase assembly protein 10 cytochrome c oxidase subunit X heme A: farnesyltransferase heme O synthase LOC691853 protoheme IX farnesyltransferase mitochondrial similar to COX10 homolog cytochrome c oxidase assembly protein heme A: farnesyltransferase; COX10 heme A:farnesyltransferase cytochrome c oxidase assembly factor; COX10 homolog, cytochrome c oxidase assembly protein, heme A: farnesyltransferase; cytochrome c oxidase assembly homolog 10; cytochrome c oxidase assembly protein; cytochrome c oxidase subunit X; heme A: farnesyltransferase; Heme O synthase; Protoheme IX farnesyltransferase, mitochondrial
Gene Aliases: COX10
UniProt ID: (Human) Q12887
Entrez Gene ID: (Human) 1352
Molecular Function: acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.