Invitrogen
Manufacturer Code:MA535479
Catalog # PIMA535479
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0960 |
Immunogen | A synthesized peptide derived from human COX1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: COX-1; COX1 Cox-1 COX3 Cox-3 Cyclooxygenase-1 P PGG/HS PCOX1 PES-1 PGG/HS PGH synthase 1 PGHS1 Pghs-1 PHS 1 PHS1 Prostaglandin G/H synthase 1 Prostaglandin H2 synthase 1 Prostaglandin-endoperoxide synthase 1 PTGHS PTGS1; cyclooxygenase 3; Cyclooxygenase-1; PGH synthase 1; PHS 1; Prostaglandin G/H synthase 1; prostaglandin G/H synthase and cyclooxygenase; Prostaglandin H2 synthase 1; Prostaglandin-endoperoxide synthase 1; prostaglandin-endoperoxide synthase 1 (prostaglandin G/H synthase and cyclooxygenase)
Gene Aliases: Cox-1; Cox-3; COX1; COX3; PCOX1; PES-1; PGG/HS; PGHS-1; PGHS1; PHS 1; PHS1; PTGHS; PTGS1
UniProt ID: (Human) P23219, (Mouse) P22437
Entrez Gene ID: (Human) 5742, (Rat) 24693, (Mouse) 19224
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.