Abnova
Manufacturer Code:PAB14841
Catalog # 89-117-300
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 2-14 at N-terminus of human COX1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C on dry atmosphere. After reconstitution with 200&muL of deionized water and concentration will be 500&mu/mL,store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of COX1.
For Research Use Only
Protein Aliases: Cytochrome c oxidase polypeptide I; Cytochrome c oxidase subunit 1; MTCO1
Gene Aliases: COI; COXI; MT-CO1; MTCO1
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.