R&D Systems
Manufacturer Code:MAB4198SP
Catalog # MAB4198SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 495222 |
Immunogen | <EM>E. coli</EM>-derived recombinant human COX-2 Ala18-Ser112 and Gln386-Leu604 Accession # P35354 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: COX-2; COX2 COX-2 COX2cyclooxygenase 2b cyclooxygenase-2 EC 1.14.99 EC 1.14.99.1 GRIPGHS hCox-2 PGG/HS PGH synthase 2 PGHS-2 PHS II PHS-2 PHS-II prostaglandin G/H synthase 2 prostaglandin G/H synthase and cyclooxygenase Prostaglandin H2 synthase 2 Prostaglandin-endoperoxide synthase 2 prostaglandin-endoperoxide synthase 2 (prostaglandin G/H synthase andcyclooxygenase) PTGS2; cyclooxygenase 2; cyclooxygenase 2b; Cyclooxygenase-2; PGH synthase 2; PHS II; Prostaglandin G/H synthase 2; Prostaglandin H2 synthase 2; Prostaglandin-endoperoxide synthase 2; prostaglandin-endoperoxide synthase 2 (prostaglandin G/H synthase and cyclooxygenase)
Gene Aliases: COX-2; COX2; GRIPGHS; hCox-2; PGG/HS; PGHS-2; PHS-2; PTGS2
UniProt ID: (Human) P35354
Entrez Gene ID: (Human) 5743
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.