Novus Biologicals
Manufacturer Code:NB1101948
Catalog # NB1101948
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This whole rabbit serum was prepared by repeated immunizations with a fusion protein corresponding to the carboxy-terminus of human COX-2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: COX-2; COX-2 COX2cyclooxygenase 2b cyclooxygenase-2 EC 1.14.99 EC 1.14.99.1 GRIPGHS hCox-2 PGG/HS PGH synthase 2 PGHS-2 PHS II PHS-2 prostaglandin G/H synthase 2 prostaglandin G/H synthase and cyclooxygenase Prostaglandin H2 synthase 2 Prostaglandin-endoperoxide synthase 2 prostaglandin-endoperoxide synthase 2 (prostaglandin G/H synthase andcyclooxygenase); cyclooxygenase 2; cyclooxygenase 2b; Cyclooxygenase-2; PGH synthase 2; PHS II; Prostaglandin G/H synthase 2; Prostaglandin H2 synthase 2; Prostaglandin-endoperoxide synthase 2; prostaglandin-endoperoxide synthase 2 (prostaglandin G/H synthase and cyclooxygenase)
Gene Aliases: COX-2; COX2; GRIPGHS; hCox-2; PGG/HS; PGHS-2; PHS-2; PTGS2
UniProt ID: (Human) P35354
Entrez Gene ID: (Human) 5743
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.