R&D Systems
Manufacturer Code:MAB3740SP
Catalog # MAB3740SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 345305 |
Immunogen | <EM>E. coli</EM>-derived recombinant human COX-1 Ala24-Leu599 Accession # P23219 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: COX-1; COX1 COX-1 COX1PGG/HS COX3 Cyclooxygenase-1 EC 1.14.99.1 PCOX1 PGH synthase 1 PGHS1 PGHS-1PHS1 PHS 1 prostaglandin G/H synthase 1 prostaglandin G/H synthase and cyclooxygenase Prostaglandin H2 synthase 1 Prostaglandin-endoperoxide synthase 1 prostaglandin-endoperoxide synthase 1 (prostaglandin G/H synthase andcyclooxygenase) PTGHS PTGS1; Cyclooxygenase-1; PGH synthase 1; Prostaglandin G/H synthase 1; Prostaglandin H2 synthase 1; Prostaglandin-endoperoxide synthase 1; prostaglandin-endoperoxide synthase 1 (prostaglandin G/H synthase and cyclooxygenase)
Gene Aliases: COX1; COX3; PCOX1; PES-1; PGG/HS; PGHS-1; PGHS1; PHS1; PTGHS; PTGS1
UniProt ID: (Human) P23219
Entrez Gene ID: (Human) 5742
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.