Novus Biologicals
Manufacturer Code:NBP298861AF594
Catalog # NB106330
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human COUP-TF II/NR2F2. |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosylation factor related protein 1; Apolipoprotein A-I regulatory protein 1; Apolipoprotein A-I regulatory protein 1 ARP-1 ARP1ADP-ribosylation factor related protein 1 chicken ovalbumin upstream promoter transcription factor 2 COUP transcription factor 2 COUP transcription factor II COUP-TF II COUP-TF2 COUPTFB COUPTFII COUP-TFII MGC117452 Nuclear receptor subfamily 2 group F member 2 nuclear receptor subfamily 2 group F member 2 SVP40 TFCOUP2apolipoprotein AI regulatory protein 1; apolipoprotein AI regulatory protein 1; ARP-1; chicken ovalbumin upstream promoter transcription factor 2; chicken ovalbumin upstream promoter-transcription factor I; COUP transcription factor 2; COUP transcription factor II; COUP-TF II; COUP-TF2; Nuclear receptor subfamily 2 group F member 2; nuclear receptor subfamily 2, group F, member 2
Gene Aliases: ARP1; CHTD4; COUPTFB; COUPTFII; NF-E3; NR2F1; NR2F2; SVP40; TFCOUP2
UniProt ID: (Human) P24468
Entrez Gene ID: (Human) 7026
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.