Proteintech
Manufacturer Code:602371IG150UL
Catalog # 50-173-6337
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4A3D9 |
Immunogen | COTL1 Fusion Protein Ag1230 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CLP Coactosin like protein COTL1; coactosin-like 1; coactosin-like F-actin binding protein 1; Coactosin-like protein
Gene Aliases: 1810074P22Rik; 2010004C08Rik; CLP; COTL1
UniProt ID: (Human) Q14019, (Mouse) Q9CQI6
Entrez Gene ID: (Pig) 100621773, (Human) 23406, (Mouse) 72042
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.