Invitrogen
Manufacturer Code:PA544327
Catalog # PIPA544327
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human COTL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: coactosin-like 1; Coactosin-like 1 (Dictyostelium) CLP FLJ43657 MGC19733; coactosin-like F-actin binding protein 1; Coactosin-like protein; coactosin-like protein {ECO:0000250|UniProtKB:Q14019}; cotl1 {ECO:0000312|EMBL:AAI58747.1, ECO:0000312|RGD:1305498}
Gene Aliases: 1810074P22Rik; 2010004C08Rik; CLP; COTL1; wu:fb08h08; zgc:63983
UniProt ID: (Bovine) Q2HJ57, (Human) Q14019, (Mouse) Q9CQI6, (Rat) B0BNA5
Entrez Gene ID: (Horse) 100070195, (Bovine) 617165, (Rabbit) 100338368, (Human) 23406, (Guinea pig) 100715923, (Mouse) 72042, (Rat) 361422, (Zebrafish) 336381
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.