Invitrogen
Manufacturer Code:PA5112931
Catalog # PIPA5112931
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Coactosin-like protein (2-142AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1810074P22Rik 2010004C08Rik BOS_16981 Clp coactosin like F-actin binding protein 1 coactosin-like 1 coactosin-like 1 (Dictyostelium) coactosin-like F-actin binding protein 1 Coactosin-like protein coactosin-like protein {ECO:0000250 Cotl1 cotl1 {ECO:0000312 EMBL:AAI58747.1 ECO:0000312 FLJ43657 LOC100338368 MGC19733 RGD:1305498} UniProtKB:Q14019} Unknown (protein for MGC:63983) wu:fb08h08 zgc:63983; coactosin-like 1; coactosin-like F-actin binding protein 1; Coactosin-like protein
Gene Aliases: 1810074P22Rik; 2010004C08Rik; CLP; COTL1
UniProt ID: (Human) Q14019, (Mouse) Q9CQI6
Entrez Gene ID: (Human) 23406, (Mouse) 72042
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.