Novus Biologicals
Manufacturer Code:NBP272086APC
Catalog # NB272086APC
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3D10 |
Immunogen | Full length recombinant protein of human CORO1B (NP_065174) produced in HEK293T cells. |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: coronin actin binding protein 1B coronin actin-binding protein 1B coronin-1B coronin-2 DKFZp762I166; coronin, actin binding protein, 1B; Coronin-1B; Coronin-2
Gene Aliases: CORO1B; CORONIN-2
UniProt ID: (Human) Q9BR76
Entrez Gene ID: (Human) 57175
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.