Novus Biologicals
Manufacturer Code:NB10061040
Catalog # NB10061040
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 450 and 489 of human Coronin actin binding protein 1B using the numbering given in entry NP_065174.1 (GeneID 57175). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: coronin actin binding protein 1B coronin actin-binding protein 1B coronin-1B coronin-2 DKFZp762I166; coronin, actin binding protein, 1B; Coronin-1B; Coronin-2
Gene Aliases: CORO1B; CORONIN-2
UniProt ID: (Human) Q9BR76
Entrez Gene ID: (Human) 57175
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.