OriGene
Manufacturer Code:CF501429
Catalog # 50-166-0446
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A5 |
Immunogen | Full length human recombit protein of human CORO1A produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CLABP CLIPINA HCORO1 IMD8 TACO p57; Clipin-A; coronin, actin binding protein, 1A; coronin-1; Coronin-1A; Coronin-like protein A; Coronin-like protein p57; TACO; Tryptophan aspartate-containing coat protein
Gene Aliases: CLABP; CLIPINA; CORO1; CORO1A; HCORO1; IMD8; p57; TACO
UniProt ID: (Human) P31146
Entrez Gene ID: (Human) 11151
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.