Invitrogen
Manufacturer Code:PA5106348
Catalog # PIPA5106348
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human COQ7 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-demethoxyubiquinone hydroxylase; 5-demethoxyubiquinone hydroxylase 5-demethoxyubiquinone hydroxylase mitochondrial CAT5 CLK1 clk-1 Coenzyme q (ubiquinone) biosynthetic enzyme (DHPB methyltransferase) 7 coenzyme Q biosynthesis protein 7 homolog coenzyme Q7 homolog ubiquinone coenzyme Q7 homolog ubiquinone (yeast) coenzyme Q7 hydroxylase COQ10D8 Coq7 COQ7 coenzyme Q 7 homolog ubiquinone demethyl-Q 7 DMQ hydroxylase placental protein KG-20 Timing protein clk-1 homolog ubiquinone biosynthesis monooxygenase COQ7 ubiquinone biosynthesis protein COQ7 homolog; 5-demethoxyubiquinone hydroxylase, mitochondrial; Coenzyme q (ubiquinone) biosynthetic enzyme (DHPB methyltransferase) 7; coenzyme Q biosynthesis protein 7 homolog; coenzyme Q7 homolog, ubiquinone; COQ7 coenzyme Q, 7 homolog ubiquinone; demethyl-Q 7; DMQ hydroxylase; placental protein KG-20; Timing protein clk-1 homolog; Ubiquinone biosynthesis monooxygenase COQ7; ubiquinone biosynthesis protein COQ7 homolog
Gene Aliases: CAT5; CLK-1; CLK1; COQ10D8; COQ7
UniProt ID: (Human) Q99807, (Mouse) P97478
Entrez Gene ID: (Human) 10229, (Rat) 25249, (Mouse) 12850
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.