Proteintech
Manufacturer Code:124811AP150UL
Catalog # 50-172-8012
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | COQ6 Fusion Protein Ag3164 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CGI 10 COQ6; Coenzyme Q10 monooxygenase 6; coenzyme Q6 homolog, monooxygenase; coenzyme Q6 monooxygenase; ubiquinone biosynthesis monooxygenase COQ6; Ubiquinone biosynthesis monooxygenase COQ6, mitochondrial
Gene Aliases: 5930427M12Rik; AW742344; CGI-10; CGI10; COQ10D6; COQ6
UniProt ID: (Human) Q9Y2Z9, (Rat) Q68FU7, (Mouse) Q8R1S0
Entrez Gene ID: (Human) 51004, (Rat) 299195, (Mouse) 217707
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.