Proteintech
Manufacturer Code:280511AP150UL
Catalog # 50-173-5811
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | COQ3 Fusion Protein Ag27740 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-polyprenyl-6-hydroxyphenol methylase; 2-polyprenyl-6-hydroxyphenyl methylase; 3,4-dihydroxy-5-hexaprenylbenzoate methyltransferase; 3-demethylubiquinol 3-O-methyltransferase; 3-demethylubiquinone-10 3-methyltransferase; 3-demethylubiquinone-9 3-methyltransferase; bA9819.1 COQ3 DHHB methyltransferase DHHB MT DHHB MTase UG0215E05; Coenzyme Q (ubiquinone); coenzyme Q3 homolog methyltransferase; coenzyme Q3 homolog, methyltransferase; coenzyme Q3 methyltransferase; DHHB methyltransferase; DHHB-MT; DHHB-MTase; dihydroxyhexaprenylbenzoate methyltransferase; hexaprenyldihydroxybenzoate methyltransferase, mitochondrial; methyltransferase COQ3; Polyprenyldihydroxybenzoate methyltransferase; Ubiquinone biosynthesis O-methyltransferase, mitochondrial
Gene Aliases: 4732433J24; bA9819.1; C77934; COQ3; DHHBMT; DHHBMTASE; UG0215E05
UniProt ID: (Human) Q9NZJ6, (Mouse) Q8BMS4
Entrez Gene ID: (Human) 51805, (Mouse) 230027
Molecular Function: methyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.