Invitrogen
Manufacturer Code:PA520539
Catalog # PIPA520539
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 15 amino acid peptide near the carboxy terminus of the human CSN8. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: COP9 (constitutive photomorphogenic) homolog, subunit 8; COP9 constitutive photomorphogenic homolog subunit 8; COP9 CSN8 SGN8 COP9 homolog COP9 signalosome complex subunit 8 JAB1-containing signalosome subunit 8 hCOP9 signalosome subunit 8; COP9 homolog; COP9 signalosome complex subunit 8; COP9 signalosome subunit 8; hCOP9; JAB1-containing signalosome subunit 8; SGN8; signalosome subunit 8
Gene Aliases: 9430009J09Rik; AA408242; COP9; COPS8; CSN8; SGN8
UniProt ID: (Human) Q99627, (Mouse) Q8VBV7, (Rat) Q6P4Z9
Entrez Gene ID: (Human) 10920, (Mouse) 108679, (Rat) 363283
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.