Abnova
Manufacturer Code:H00010920K
Catalog # 89-210-768
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human COPS8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human COPS8 peptide using ARM Technology.
For Research Use Only
Protein Aliases: COP9 constitutive photomorphogenic homolog subunit 8; COP9 CSN8 MGC1297 MGC43256 SGN8; COP9 homolog; COP9 signalosome complex subunit 8; hCOP9; JAB1-containing signalosome subunit 8; SGN8; signalosome subunit 8
Gene Aliases: COP9; COPS8; CSN8; SGN8
UniProt ID: (Human) Q99627
Entrez Gene ID: (Human) 10920
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.