Abnova
Manufacturer Code:MAB12626
Catalog # 89-933-988
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 8H8.5 |
Immunogen | Recombinant protein corresponding to mouse Cops5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against mouse Cops5.
For Research Use Only
Protein Aliases: AI303502; COP9 complex S5; COP9 signalosome complex subunit 5; CSN5; Jab1; Jun activation domain-binding protein 1; Jun coactivator; Kip1 C-terminus-interacting protein 2; Mov34; SGN5
Gene Aliases: AI303502; Cops5; CSN5; Jab1; Kic2; Mov34; Sgn5
UniProt ID: (Mouse) O35864
Entrez Gene ID: (Mouse) 26754
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.