Invitrogen
Manufacturer Code:PA583634
Catalog # PIPA583634
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human COPS4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: COP9 constitutive photomorphogenic homolog subunit 4; COP9 constitutive photomorphogenic-like protein subunit 4; COP9 signalosome complex subunit 4; CSN4; JAB1-containing signalosome subunit 4; SGN4; signalosome subunit 4; testis tissue sperm-binding protein Li 42a
Gene Aliases: COPS4; CSN4
UniProt ID: (Human) B3KN88
Entrez Gene ID: (Human) 51138
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.