Proteintech
Manufacturer Code:104641AP150UL
Catalog # 50-172-6326
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | COPS4 Fusion Protein Ag0735 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: COP9 (constitutive photomorphogenic) homolog, subunit 4; COP9 (constitutive photomorphogenic), subunit 4; COP9 complex S4; COP9 constitutive photomorphogenic homolog subunit 4; COP9 constitutive photomorphogenic-like protein subunit 4; COP9 signalosome complex subunit 4; COPS4 CSN4 SGN4 Signalosome subunit 4; JAB1-containing signalosome subunit 4; SGN4; signalosome subunit 4; testis tissue sperm-binding protein Li 42a
Gene Aliases: AW208976; COPS4; CSN4; D5Ertd774e
UniProt ID: (Human) B3KN88, (Mouse) O88544, (Rat) Q68FS2
Entrez Gene ID: (Human) 51138, (Mouse) 26891, (Rat) 360915
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.