Proteintech
Manufacturer Code:123931AP150UL
Catalog # 50-172-7952
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | COPG Fusion Protein Ag3070 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: coat protein gamma-cop; coatomer protein complex, subunit gamma 1; coatomer subunit gamma; Coatomer subunit gamma COPG COPG1 Gamma coat protein Gamma COP; Coatomer subunit gamma-1; Gamma-1-coat protein; Gamma-1-COP; gamma-coat protein; gamma-COP
Gene Aliases: AU019265; BC056168; COPG; COPG1; D6Ertd71e; D6Wsu16e
UniProt ID: (Human) Q9Y678, (Mouse) Q9QZE5, (Rat) Q4AEF8
Entrez Gene ID: (Human) 22820, (Mouse) 54161, (Rat) 297428
Molecular Function:
membrane traffic protein
vesicle coat protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.