Novus Biologicals
Manufacturer Code:NB100363
Catalog # NB100363
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human COP9 constitutive photomorphogenic homolog subunit 2 using the numbering given in entry NP_004227.1 (GeneID 9318). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ALIEN Alien homolog COP9 constitutive photomorphogenic homolog subunit 2 (Arabidopsis) COP9 signalosome complex subunit 2 CSN2JAB1-containing signalosome subunit 2 SGN2TR-interacting protein 15 Signalosome subunit 2 thyroid receptor interacting protein 15 Thyroid receptor-interacting protein 15 TRIP15TRIP-15; Alien homolog; COP9 constitutive photomorphogenic homolog subunit 2; COP9 signalosome complex subunit 2; JAB1-containing signalosome subunit 2; SGN2; signalosome subunit 2; thyroid receptor interacting protein 15; Thyroid receptor-interacting protein 15; TR-interacting protein 15; TRIP-15
Gene Aliases: ALIEN; COPS2; CSN2; SGN2; TRIP15
UniProt ID: (Human) P61201
Entrez Gene ID: (Human) 9318
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.