Novus Biologicals
Manufacturer Code:NBP185435
Catalog # NBP185435
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:KYVLHMIEDGEIFASINQKDGMVSFHDNPEKYNNPAMLHNIDQEMLKCIELDERLKAMDQEITVNPQFVQKSMGSQEDDSGNKPSSYS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: COP9 complex subunit 3; COP9 constitutive photomorphogenic homolog subunit 3; COP9 constitutive photomorphogenic homolog subunit 3 (Arabidopsis) COP9 signalosome complex subunit 3 CSN3COP9 complex subunit 3 JAB1-containing signalosome subunit 3 SGN3COP9 (constitutive photomorphogenic Arabidopsis homolog) subunit 3 Signalosome subunit 3; COP9 signalosome complex subunit 3; JAB1-containing signalosome subunit 3; SGN3; signalosome subunit 3
Gene Aliases: COPS3; CSN3; SGN3
UniProt ID: (Human) Q9UNS2
Entrez Gene ID: (Human) 8533
Molecular Function:
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.