Invitrogen
Manufacturer Code:MA535515
Catalog # PIMA535515
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1006 |
Immunogen | A synthesized peptide derived from human COMT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Catechol O-methyltransferase; Catechol O-methyltransferase catechol-O-methyltransferase Comt Comt1 D16Wsu103e D330014B15Rik HEL-S-98n OTTHUMP00000197749 OTTHUMP00000197750; catechol-O-methyltransferase isoform; catecholamine-O-methyltransferase; epididymis secretory sperm binding protein Li 98n; testicular tissue protein Li 42
Gene Aliases: COMT; HEL-S-98n
UniProt ID: (Human) P21964, (Rat) P22734
Entrez Gene ID: (Human) 1312, (Rat) 24267
Molecular Function:
methyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.