Proteintech
Manufacturer Code:147541AP150UL
Catalog # 50-172-9698
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | COMT Fusion Protein Ag6502 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: catechol O methyltransferase COMT; Catechol O-methyltransferase; catechol-O-methyltransferase 1; catechol-O-methyltransferase isoform; catecholamine-O-methyltransferase; epididymis secretory sperm binding protein Li 98n; testicular tissue protein Li 42
Gene Aliases: COMT; Comt1; D16Wsu103e; D330014B15Rik; HEL-S-98n
UniProt ID: (Human) P21964, (Mouse) O88587, (Rat) P22734
Entrez Gene ID: (Human) 1312, (Mouse) 12846, (Rat) 24267
Molecular Function:
methyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.