Invitrogen
Manufacturer Code:PA567956
Catalog # PIPA567956
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human COLQ. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetylcholinesterase collagenic tail peptide; Acetylcholinesterase-associated collagen; AChE Q subunit; collagen-like tail subunit (single strand of homotrimer) of asymmetric acetylcholinesterase; collagen-like tail subunit of asymmetric acetylcholinesterase; collagenic tail of endplate acetylcholinesterase; EAD AChE Q subunit acetylcholinesterase collagenic tail peptide acetylcholinesterase-associated collagen collagenic tail of endplate acetylcholinesterase single strand of homotrimeric collagen-like tail subunit of asymmetric acetylcholinesterase; single strand of homotrimeric collagen-like tail subunit of asymmetric acetylcholinesterase
Gene Aliases: A130034K24Rik; CMS5; COLQ; EAD
UniProt ID: (Human) Q6YH20, (Mouse) O35348, (Rat) O35167
Entrez Gene ID: (Human) 8292, (Mouse) 382864, (Rat) 29755
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.