Novus Biologicals
Manufacturer Code:NBP2847140.1ML
Catalog # NB032642
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C terminal region of human COLQ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetylcholinesterase collagenic tail peptide; acetylcholinesterase collagenic tail peptide AChE Q subunit collagenic tail of endplate acetylcholinesterase collagen-like tail subunit (single strand of homotrimer) of asymmetricacetylcholinesterase EADAcetylcholinesterase-associated collagen FLJ55041 single strand of homotrimeric collagen-like tail subunit of asymmetricacetylcholinesterase; Acetylcholinesterase-associated collagen; AChE Q subunit; collagen-like tail subunit (single strand of homotrimer) of asymmetric acetylcholinesterase; collagenic tail of endplate acetylcholinesterase; single strand of homotrimeric collagen-like tail subunit of asymmetric acetylcholinesterase
Gene Aliases: CMS5; COLQ; EAD
UniProt ID: (Human) Q6YH20
Entrez Gene ID: (Human) 8292
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.