Abnova
Manufacturer Code:H00008292A01
Catalog # 89-105-828
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 356 a.a. to 455 a.a) partial recombinant protein with GST tag.,COLQ (NP_005668 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant COLQ.
For Research Use Only
Protein Aliases: Acetylcholinesterase collagenic tail peptide; Acetylcholinesterase-associated collagen; AChE Q subunit; collagen-like tail subunit (single strand of homotrimer) of asymmetric acetylcholinesterase; collagenic tail of endplate acetylcholinesterase; EAD; single strand of homotrimeric collagen-like tail subunit of asymmetric acetylcholinesterase
Gene Aliases: CMS5; COLQ; EAD
UniProt ID: (Human) Q6YH20
Entrez Gene ID: (Human) 8292
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.