Proteintech
Manufacturer Code:152691AP150UL
Catalog # 50-173-0105
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | COLEC11 Fusion Protein Ag7374 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CL K1 CL K1 I CL K1 II CL K1 IIa CL K1 IIb CLK1 COLEC11 Collectin 11 Collectin kidney protein 1 collectin sub family member 11; CL-K1; Collectin K1; Collectin kidney protein 1; collectin sub-family member 11; Collectin-11
Gene Aliases: 1010001H16Rik; 3MC2; CL-K1-I; CL-K1-II; CL-K1-IIa; CL-K1-IIb; CLK1; COLEC11; UNQ596/PRO1182; wu:fb82e09; zgc:103572
UniProt ID: (Human) Q9BWP8, (Mouse) Q3SXB8, (Zebrafish) Q5U3G1
Entrez Gene ID: (Human) 78989, (Mouse) 71693, (Rat) 366588, (Zebrafish) 492459
Molecular Function: defense/immunity protein surfactant
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.