Novus Biologicals
Manufacturer Code:NBP292796100UL
Catalog # NB027458
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 20-270 of human COL9A1 (NP_001842.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha-1 polypeptide collagen type IX alpha 1; alpha-1(IX) collagen chain; cartilage-specific short collagen; Collagen alpha-1(IX) chain; collagen IX, alpha-1 polypeptide; collagen, type IX, alpha 1
Gene Aliases: COL9A1; DJ149L1.1.2; EDM6; MED; STL4
UniProt ID: (Human) Q9H151
Entrez Gene ID: (Human) 1297
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.