Invitrogen
Manufacturer Code:PA518824
Catalog # PIPA518824
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (DHDSAPLNNRC) corresponding to the internal amino acids of COL4A3BP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles,Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Ceramide transfer protein; ceramide transport protein; CERT; COL4A3BP collagen type IV alpha 3 (Goodpasture antigen) binding protein CERT CERTL FLJ20597 GPBP STARD11 Goodpasture antigen-binding protein START domain containing 11 alpha 3 type IV collagen binding protein ceramide transporter; Collagen type IV alpha-3-binding protein; Goodpasture antigen-binding protein; GPBP; procollagen, type IV, alpha 3 (Goodpasture antigen) binding protein; StAR-related lipid transfer protein 11; StARD11; START domain-containing protein 11
Gene Aliases: 2810404O15Rik; 9230101K08Rik; AU016711; CERT; Cert1; Col4a3bp; GPBP; Stard11
UniProt ID: (Mouse) Q9EQG9
Entrez Gene ID: (Mouse) 68018
Molecular Function: kinase protein kinase transfer/carrier protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.